Assay Details
Gene Symbol: Src64B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014561.2 | NP_001014561.1 | 6 - 7 | 2071 | 62 | |
NM_001014562.3 | NP_001014562.1 | 5 - 6 | 1823 | 62 | ||
NM_001014563.3 | NP_001014563.1 | 7 - 8 | 2212 | 62 | ||
NM_001014564.3 | NP_001014564.1 | 7 - 8 | 1973 | 62 | ||
NM_001202122.1 | NP_001189051.1 | 6 - 7 | 1766 | 62 | ||
NM_001259699.2 | NP_001246628.1 | 7 - 8 | 1973 | 62 | ||
NM_001259700.2 | NP_001246629.1 | 7 - 8 | 1973 | 62 | ||
NM_001300008.1 | NP_001286937.1 | 7 - 8 | 2041 | 62 | ||
NM_080195.4 | NP_524934.2 | 7 - 8 | 2038 | 62 | ||
GenBank mRNA | AY051781.1 | - | 7 - 8 | 2015 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
