Assay Details
| Gene Symbol: cmet | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273452.1 | NP_001260381.1 | 4 - 5 | 1669 | 108 | |
| NM_080254.3 | NP_524993.3 | 4 - 5 | 1669 | 108 | ||
More Information
Set Membership: |
|

cmet
Genomic Map