Assay Details
| Gene Symbol: Raf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103397.3 | NP_001096867.3 | 4 - 5 | 2712 | 70 | |
| NM_080308.4 | NP_525047.1 | 2 - 3 | 1712 | 70 | ||
| GenBank mRNA | AY089490.1 | - | 2 - 3 | 1667 | 70 | |
More Information
Set Membership: |
|

Raf
Genomic Map