Assay Details
Gene Symbol: Raf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103397.3 | NP_001096867.3 | 5 - 6 | 2856 | 99 | |
NM_080308.4 | NP_525047.1 | 3 - 4 | 1856 | 99 | ||
GenBank mRNA | AY089490.1 | - | 3 - 4 | 1811 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
