Assay Details
| Gene Symbol: Fas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080327.3 | NP_525066.1 | 8 - 9 | 2834 | 51 | |
| GenBank mRNA | M77166.1 | - | 8 - 9 | 2775 | 51 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fas2| Gene Symbol: Fas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080327.3 | NP_525066.1 | 8 - 9 | 2834 | 51 | |
| GenBank mRNA | M77166.1 | - | 8 - 9 | 2775 | 51 | |
Set Membership: |
|