Assay Details
| Gene Symbol: betaggt-I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273132.1 | NP_001260061.1 | 2 - 3 | 218 | 54 | |
| NM_080361.3 | NP_525100.2 | 2 - 3 | 218 | 54 | ||
| GenBank mRNA | AY119246.1 | - | 2 - 3 | 187 | 54 | |
More Information
Set Membership: |
|

betaggt-I
Genomic Map