Assay Details
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080378.4 | NP_525117.2 | 4 - 5 | 1656 | 79 | |
GenBank mRNA | AB176552.1 | - | 4 - 5 | 1095 | 79 | |
BT032838.1 | - | 4 - 5 | 1612 | 79 | ||
BT124781.1 | - | 5 - 6 | 2487 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
