Assay Details
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080378.4 | NP_525117.2 | 5 - 6 | 1738 | 73 | |
GenBank mRNA | AB176552.1 | - | 5 - 6 | 1177 | 73 | |
BT032838.1 | - | 5 - 6 | 1694 | 73 | ||
BT124781.1 | - | 6 - 7 | 2569 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
