Assay Details
Gene Symbol: Mgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080378.4 | NP_525117.2 | 6 - 7 | 1827 | 89 | |
GenBank mRNA | AB176552.1 | - | 6 - 7 | 1266 | 89 | |
BT032838.1 | - | 6 - 7 | 1783 | 89 | ||
BT124781.1 | - | 7 - 8 | 2658 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
