Assay Details
| Gene Symbol: cype | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258956.2 | NP_001245885.1 | 2 - 3 | 232 | 81 | |
| NM_001258957.2 | NP_001245886.1 | 2 - 3 | 230 | 81 | ||
| NM_080526.3 | NP_536787.1 | 2 - 3 | 230 | 81 | ||
| GenBank mRNA | KX532000.1 | - | 1 - 2 | 166 | 81 | |
More Information
Set Membership: |
|

cype
Genomic Map