Assay Details
| Gene Symbol: Gss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298465.1 | NP_001285394.1 | 2 - 3 | 537 | 85 | |
| Gene Symbol: Gss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298466.1 | NP_001285395.1 | 2 - 3 | 537 | 85 | |
More Information
Set Membership: |
|

Gss1
Genomic Map