Assay Details
| Gene Symbol: Ggt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298479.1 | NP_001285408.1 | 2 - 3 | 1507 | 85 | |
| NM_133075.3 | NP_573303.1 | 2 - 3 | 1691 | 85 | ||
| GenBank mRNA | AY058701.1 | - | 2 - 3 | 1524 | 85 | |
More Information
Set Membership: |
|

Ggt-1
Genomic Map