Assay Details
Gene Symbol: Zasp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032250.3 | NP_001027421.1 | 11 - 12 | 1916 | 145 | |
NM_001043086.3 | NP_001036551.1 | 12 - 13 | 2246 | 145 | ||
NM_001274082.1 | NP_001261011.1 | 11 - 12 | 1742 | 145 | ||
NM_001316422.1 | NP_001303351.1 | 4 - 5 | 981 | 145 | ||
NM_145757.3 | NP_665700.2 | 12 - 13 | 2878 | 145 | ||
GenBank mRNA | BT001752.1 | - | 11 - 12 | 1913 | 145 | |
BT022171.2 | - | 12 - 13 | 2226 | 145 | ||
JN034424.1 | - | 11 - 12 | 1738 | 145 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
