Assay Details
| Gene Symbol: Ank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169348.2 | NP_001162819.1 | 5 - 6 | 1502 | 93 | |
| NM_175926.3 | NP_787122.1 | 4 - 5 | 1460 | 93 | ||
| NM_175927.3 | NP_787123.1 | 5 - 6 | 1535 | 93 | ||
| NM_175928.3 | NP_787124.1 | 5 - 6 | 1650 | 93 | ||
| GenBank mRNA | BT031123.1 | - | 4 - 5 | 1455 | 93 | |
More Information
Set Membership: |
|

Ank
Genomic Map