Assay Details
Gene Symbol: Ank | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169348.2 | NP_001162819.1 | 8 - 9 | 2862 | 62 | |
NM_175926.3 | NP_787122.1 | 7 - 8 | 2820 | 62 | ||
NM_175927.3 | NP_787123.1 | 8 - 9 | 2895 | 62 | ||
NM_175928.3 | NP_787124.1 | 8 - 9 | 3010 | 62 | ||
GenBank mRNA | BT031123.1 | - | 7 - 8 | 2815 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
