Assay Details
| Gene Symbol: mRpL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273099.1 | NP_001260028.1 | 2 - 3 | 287 | 64 | |
| NM_001298673.1 | NP_001285602.1 | 2 - 3 | 323 | 64 | ||
| NM_175957.2 | NP_787971.1 | 2 - 3 | 287 | 64 | ||
More Information
Set Membership: |
|

mRpL27
Genomic Map