Assay Details
Gene Symbol: TER94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103779.2 | NP_001097249.1 | 2 - 3 | 288 | 78 | |
NM_001103780.2 | NP_001097250.1 | 2 - 3 | 355 | 78 | ||
NM_001299332.1 | NP_001286261.1 | 2 - 3 | 288 | 78 | ||
NM_058021.4 | NP_477369.1 | 2 - 3 | 246 | 78 | ||
GenBank mRNA | AF132553.1 | - | 2 - 3 | 228 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001103780.2 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
