Assay Details
| Gene Symbol: Traf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103610.2 | NP_001097080.2 | 3 | 2509 | 146 | |
| NM_058068.4 | NP_477416.1 | 2 | 2195 | 146 | ||
| GenBank mRNA | AF119794.1 | - | 2 | 2014 | 146 | |
| AY094792.1 | - | 2 | 2174 | 146 | ||
More Information
Set Membership: |
|

Traf4
Genomic Map