Assay Details
| Gene Symbol: Eno | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058073.5 | NP_477421.1 | 1 - 2 | 71 | 90 | |
| NM_164431.3 | NP_722721.1 | 3 - 4 | 500 | 90 | ||
| NM_164432.3 | NP_722722.1 | 3 - 4 | 574 | 90 | ||
| NM_164433.3 | NP_722723.1 | 3 - 4 | 515 | 90 | ||
| NM_164434.3 | NP_722724.1 | 3 - 4 | 775 | 90 | ||
| GenBank mRNA | BT058002.1 | - | 3 - 4 | 493 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_058073.5 transcripts.
Set Membership: |
|

Eno
Genomic Map