Assay Details
Gene Symbol: esc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058083.4 | NP_477431.1 | 3 - 4 | 535 | 72 | |
GenBank mRNA | AY069796.1 | - | 3 - 4 | 507 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
