Assay Details
Gene Symbol: cass | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058106.5 | NP_477454.1 | 3 - 4 | 933 | 76 | |
GenBank mRNA | AF160921.1 | - | 3 - 4 | 878 | 76 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
