Assay Details
| Gene Symbol: beat-Ia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299046.1 | NP_001285975.1 | 3 - 4 | 1040 | 72 | |
| NM_058160.4 | NP_477508.1 | 3 - 4 | 1040 | 72 | ||
| GenBank mRNA | BT150361.1 | - | 3 - 4 | 1008 | 72 | |
More Information
Set Membership: |
|

beat-Ia
Genomic Map