Assay Details
Gene Symbol: beat-Ia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299046.1 | NP_001285975.1 | 6 - 7 | 1433 | 97 | |
NM_058160.4 | NP_477508.1 | 6 - 7 | 1433 | 97 | ||
GenBank mRNA | U67057.1 | - | 6 - 7 | 1376 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
