Assay Details
Gene Symbol: beat-Ia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058160.4 | NP_477508.1 | 7 - 8 | 1630 | 76 | |
GenBank mRNA | BT150361.1 | - | 7 - 8 | 1598 | 76 | |
U67057.1 | - | 7 - 8 | 1573 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
