Assay Details
| Gene Symbol: Naa15-16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298510.1 | NP_001285439.1 | 3 - 4 | 1968 | 60 | |
| NM_133156.2 | NP_573384.1 | 3 - 4 | 1693 | 60 | ||
| GenBank mRNA | BT014631.1 | - | 3 - 4 | 1647 | 60 | |
More Information
Set Membership: |
|

Naa15-16
Genomic Map