Assay Details
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103574.3 | NP_001097044.2 | 4 - 5 | 844 | 61 | |
NM_001103575.2 | NP_001097045.1 | 3 - 4 | 736 | 61 | ||
NM_001258876.2 | NP_001245805.1 | 3 - 4 | 973 | 61 | ||
NM_001258877.2 | NP_001245806.1 | 3 - 4 | 722 | 61 | ||
NM_001258878.2 | NP_001245807.1 | 3 - 4 | 753 | 61 | ||
NM_001258879.2 | NP_001245808.1 | 4 - 5 | 1225 | 61 | ||
NM_164353.2 | NP_722589.1 | 3 - 4 | 742 | 61 | ||
NM_164354.3 | NP_722590.2 | 3 - 4 | 742 | 61 | ||
GenBank mRNA | BT057995.1 | - | 3 - 4 | 717 | 61 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
