Assay Details
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103575.2 | NP_001097045.1 | 4 - 5 | 923 | 122 | |
NM_001258878.2 | NP_001245807.1 | 4 - 5 | 940 | 122 | ||
NM_001258879.2 | NP_001245808.1 | 5 - 6 | 1412 | 122 | ||
NM_164353.2 | NP_722589.1 | 4 - 5 | 929 | 122 | ||
NM_164354.3 | NP_722590.2 | 4 - 5 | 929 | 122 | ||
GenBank mRNA | BT057995.1 | - | 4 - 5 | 904 | 122 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
