Assay Details
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103574.3 | NP_001097044.2 | 7 - 8 | 4083 | 79 | |
NM_001103575.2 | NP_001097045.1 | 5 - 6 | 1422 | 79 | ||
NM_001258877.2 | NP_001245806.1 | 6 - 7 | 3661 | 79 | ||
NM_001258878.2 | NP_001245807.1 | 5 - 6 | 1439 | 79 | ||
NM_001258879.2 | NP_001245808.1 | 6 - 7 | 1911 | 79 | ||
NM_164353.2 | NP_722589.1 | 5 - 6 | 1428 | 79 | ||
NM_164354.3 | NP_722590.2 | 5 - 6 | 1428 | 79 | ||
GenBank mRNA | BT057995.1 | - | 5 - 6 | 1403 | 79 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
