Assay Details
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103574.3 | NP_001097044.2 | 11 - 12 | 4850 | 62 | |
NM_001258876.2 | NP_001245805.1 | 7 - 8 | 3995 | 62 | ||
NM_001258877.2 | NP_001245806.1 | 10 - 11 | 4428 | 62 | ||
NM_001258878.2 | NP_001245807.1 | 8 - 9 | 1762 | 62 | ||
NM_001258879.2 | NP_001245808.1 | 10 - 11 | 5111 | 62 | ||
NM_164353.2 | NP_722589.1 | 9 - 10 | 2195 | 62 | ||
NM_164354.3 | NP_722590.2 | 10 - 11 | 5072 | 62 | ||
GenBank mRNA | BT057995.1 | - | 9 - 10 | 2170 | 62 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
