Assay Details
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103574.3 | NP_001097044.2 | 5 - 6 | 1041 | 132 | |
NM_001258876.2 | NP_001245805.1 | 4 - 5 | 1170 | 132 | ||
NM_001258877.2 | NP_001245806.1 | 4 - 5 | 919 | 132 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
