Assay Details
Gene Symbol: Cda5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258879.2 | NP_001245808.1 | 8 - 9 | 4831 | 60 | |
NM_164354.3 | NP_722590.2 | 8 - 9 | 4792 | 60 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
