Assay Details
Gene Symbol: Gs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057222.4 | NP_476570.1 | 2 | 2103 | 140 | |
NM_164367.2 | NP_722606.1 | 1 | 2323 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
