Assay Details
Gene Symbol: Nnf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164388.3 | NP_722652.1 | 3 - 4 | 506 | 95 | |
GenBank mRNA | BT015998.1 | - | 3 - 4 | 497 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
