Assay Details
| Gene Symbol: Kebab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164451.2 | NP_722756.1 | 2 - 3 | 461 | 69 | |
| GenBank mRNA | AY051677.1 | - | 2 - 3 | 430 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kebab| Gene Symbol: Kebab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164451.2 | NP_722756.1 | 2 - 3 | 461 | 69 | |
| GenBank mRNA | AY051677.1 | - | 2 - 3 | 430 | 69 | |
Set Membership: |
|