Assay Details
| Gene Symbol: Kebab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164451.2 | NP_722756.1 | 4 - 5 | 1818 | 90 | |
| GenBank mRNA | AY051677.1 | - | 4 - 5 | 1787 | 90 | |
More Information
Set Membership: |
|

Kebab
Genomic Map