Assay Details
Gene Symbol: Reph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164561.3 | NP_722945.2 | 1 - 2 | 1141 | 75 | |
GenBank mRNA | AY119477.1 | - | 1 - 2 | 1125 | 75 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
