Assay Details
| Gene Symbol: CG31648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164644.3 | NP_723086.1 | 1 | 788 | 79 | |
| GenBank mRNA | BT011370.1 | - | 1 | 781 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CG31648| Gene Symbol: CG31648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164644.3 | NP_723086.1 | 1 | 788 | 79 | |
| GenBank mRNA | BT011370.1 | - | 1 | 781 | 79 | |
Set Membership: |
|