Assay Details
Gene Symbol: Hel25E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273171.1 | NP_001260100.1 | 4 - 5 | 797 | 66 | |
NM_164646.3 | NP_723089.1 | 4 - 5 | 797 | 66 | ||
NM_164647.1 | NP_723090.1 | 3 - 4 | 687 | 66 | ||
NM_164648.2 | NP_723091.1 | 4 - 5 | 790 | 66 | ||
GenBank mRNA | AY118921.1 | - | 4 - 5 | 752 | 66 | |
X79802.1 | - | 768 | 66 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
