Assay Details
Gene Symbol: Hel25E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_164646.3 | NP_723089.1 | 5 - 6 | 917 | 65 | |
NM_164647.1 | NP_723090.1 | 4 - 5 | 807 | 65 | ||
NM_164648.2 | NP_723091.1 | 5 - 6 | 910 | 65 | ||
GenBank mRNA | AY118921.1 | - | 5 - 6 | 872 | 65 | |
X79802.1 | - | 888 | 65 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
