Assay Details
| Gene Symbol: Hel25E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273171.1 | NP_001260100.1 | 6 - 7 | 2119 | 69 | |
| NM_164646.3 | NP_723089.1 | 7 - 8 | 1448 | 69 | ||
| NM_164647.1 | NP_723090.1 | 6 - 7 | 1338 | 69 | ||
| NM_164648.2 | NP_723091.1 | 7 - 8 | 1441 | 69 | ||
| GenBank mRNA | AY118921.1 | - | 7 - 8 | 1403 | 69 | |
| X79802.1 | - | 1419 | 69 | |||
More Information
Set Membership: |
|

Hel25E
Genomic Map