Assay Details
Gene Symbol: Gyc32E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103678.2 | NP_001097148.1 | 11 - 12 | 1595 | 76 | |
NM_001298905.1 | NP_001285834.1 | 10 - 11 | 1487 | 76 | ||
NM_176013.2 | NP_788027.1 | 10 - 11 | 1511 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
