Assay Details
| Gene Symbol: Gyc32E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103678.2 | NP_001097148.1 | 22 - 23 | 3430 | 91 | |
| NM_001298905.1 | NP_001285834.1 | 21 - 22 | 3322 | 91 | ||
| NM_176013.2 | NP_788027.1 | 21 - 22 | 3346 | 91 | ||
More Information
Set Membership: |
|

Gyc32E
Genomic Map