Assay Details
| Gene Symbol: Ance-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038815.2 | NP_001033904.1 | 6 - 7 | 2263 | 97 | |
| NM_001298988.1 | NP_001285917.1 | 6 - 7 | 2263 | 97 | ||
| GenBank mRNA | AY075532.1 | - | 6 - 7 | 2263 | 97 | |
More Information
Set Membership: |
|

Ance-3
Genomic Map