Assay Details
| Gene Symbol: NimC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298993.1 | NP_001285922.1 | 1 - 2 | 145 | 59 | |
| NM_176033.3 | NP_788047.1 | 1 - 2 | 145 | 59 | ||
More Information
Set Membership: |
|

NimC2
Genomic Map