Assay Details
Gene Symbol: NimC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298993.1 | NP_001285922.1 | 2 - 3 | 252 | 63 | |
NM_176033.3 | NP_788047.1 | 2 - 3 | 252 | 63 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
