Assay Details
Gene Symbol: MED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176220.2 | NP_788400.1 | 3 - 4 | 695 | 71 |
Gene Symbol: CG42518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169716.2 | NP_001163187.1 | 3 - 4 | 695 | 71 | |
NM_001169717.1 | NP_001163188.1 | 4 - 5 | 649 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
