Assay Details
Gene Symbol: MED9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176220.2 | NP_788400.1 | 2 - 3 | 331 | 82 | |
GenBank mRNA | KX531396.1 | - | 2 - 3 | 269 | 82 |
Gene Symbol: CG42518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169716.2 | NP_001163187.1 | 2 - 3 | 331 | 82 | |
NM_001169717.1 | NP_001163188.1 | 3 - 4 | 285 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001169716.2, NM_001169717.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
