Assay Details
Gene Symbol: CG33057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300051.1 | NP_001286980.1 | 2 | 2981 | 99 | |
NM_176299.3 | NP_788477.1 | 1 | 3033 | 99 | ||
GenBank mRNA | AY060662.1 | - | 1 | 3028 | 99 |
Gene Symbol: mkg-p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300052.1 | NP_001286981.1 | 2 | 2981 | 99 | |
NM_139962.4 | NP_648219.1 | 2 | 2260 | 99 | ||
NM_168272.2 | NP_729375.1 | 3 | 2278 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
