Assay Details
Gene Symbol: kay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032407.3 | NP_001027579.1 | 1 - 2 | 936 | 68 | |
GenBank mRNA | AF238310.1 | - | 998 | 68 | ||
AY574373.1 | - | 2 - 3 | 787 | 68 | ||
BT088433.1 | - | 1 - 2 | 841 | 68 | ||
BT099528.1 | - | 1 - 2 | 841 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
