Assay Details
Gene Symbol: kay | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032405.4 | NP_001027577.2 | 2 - 3 | 2252 | 87 | |
NM_001032406.2 | NP_001027578.1 | 2 - 3 | 872 | 87 | ||
NM_001032407.3 | NP_001027579.1 | 2 - 3 | 1263 | 87 | ||
NM_001032408.3 | NP_001027580.1 | 2 - 3 | 1926 | 87 | ||
NM_001300662.1 | NP_001287591.1 | 3 - 4 | 1416 | 87 | ||
GenBank mRNA | AF238310.1 | - | 1325 | 87 | ||
AY069805.1 | - | 2 - 3 | 849 | 87 | ||
AY574373.1 | - | 3 - 4 | 1114 | 87 | ||
BT088433.1 | - | 2 - 3 | 1168 | 87 | ||
BT099528.1 | - | 2 - 3 | 1168 | 87 | ||
DQ858476.1 | - | 2 - 3 | 870 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
